accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 43119-28-4

IUPAC Name: (3aR,6aS)-2H,3H,3aH,6H,6aH-cyclopenta[b]furan-2-one
Synonyms: (3aR,6aS)-3H,3aH,6H,6aH-cyclopenta[b]furan-2-one
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: RYBPGUMSFWGGLP-WDSKDSINSA-N
SMILES: O=C1C[C@@H]2C=CC[C@@H]2O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results