Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,5R)-(-)-cis-2-Oxabicyclo[3.3.0]oct-6-en-3-one 97%, Thermo Scientific™

Catalog No. AC233490010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC233490010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC233490010 Supplier Thermo Scientific Chemicals Supplier No. 233490010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43119-28-4
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00010402
InChI Key RYBPGUMSFWGGLP-WDSKDSINSA-N
Synonym 3ar,6as-3,3a,6,6a-tetrahydro-2h-cyclopenta b furan-2-one, 1s,5r-2-oxabicyclo 3.3.0 oct-6-en-3-one, 2h-cyclopenta b furan-2-one, 3,3a,6,6a-tetrahydro-, 3ar,6as, 1s,5r---2-oxabicyclo 3.3.0 oct-6-en-3-one, 3ar,6as-3,3a,6,6a-tetrahydrocyclopenta b furan-2-one, 3as,6ar-rel-3,3a,6,6a-tetrahydro-2h-cyclopenta b furan-2-one, 2h-cyclopenta b furan-2-one,3,3a,6,6a-tetrahydro-, 3ar,6as, 3ar,6as-3h,3ah,6h,6ah-cyclopenta b furan-2-one, 1s,5r---cis-2-oxabicyclo 3.3.0 oct-6-en-3-one, 1s,5r-oxabicyclooctenone
PubChem CID 643405
IUPAC Name (3aR,6aS)-2H,3H,3aH,6H,6aH-cyclopenta[b]furan-2-one
SMILES O=C1C[C@@H]2C=CC[C@@H]2O1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Beige to White or Brown
Infrared Spectrum Authentic
Quantity 1 g
Specific Rotation (25°C, 589nm) (c=1, MeOH) < -97°
Formula Weight 124.14
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material (1S, 5R)-(-)-cis-2-Oxabicyclo[3.3.0]oct-6-en-3-one, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.