accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 43141-69-1

IUPAC Name: 3-(dibutylamino)phenol
Synonyms: 3-(dibutylamino)phenol
Molecular Weight (g/mol): 221.34
Molecular Formula: C14H23NO
InChi Key: KHSTZMGCKHBFJX-UHFFFAOYSA-N
SMILES: CCCCN(CCCC)C1=CC(O)=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results