Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dibutyl-3-aminophenol 95.0+%, TCI America™
SDP

Catalog No. D2138500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2138500G 500 g
D213825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2138500G Supplier TCI America Supplier No. D2138500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 43141-69-1
Molecular Formula C14H23NO
Molecular Weight (g/mol) 221.34
MDL Number MFCD00134668
InChI Key KHSTZMGCKHBFJX-UHFFFAOYSA-N
PubChem CID 601234
IUPAC Name 3-(dibutylamino)phenol
SMILES CCCCN(CCCC)C1=CC(O)=CC=C1

Specifications

Boiling Point 176°C
Quantity 500 g
Formula Weight 221.34
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Dibutyl-3-aminophenol
EINECSNumber (3)-3804
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.