CAS RN 4316-35-2
IUPAC Name:
(1,1-dimethoxyethyl)benzene
Synonyms:
(1,1-dimethoxyethyl)benzene
Molecular Weight (g/mol):
166.22
Molecular Formula:
C10H14O2
InChi Key:
XKSUVRWJZCEYQQ-UHFFFAOYSA-N
SMILES:
COC(C)(OC)C1=CC=CC=C1
1
–
1
of
1
results
1
Sigma Aldrich (1,1-Dimethoxyethyl)benzene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.