accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4316-53-4

IUPAC Name: 4-methyl-N,N-diphenylaniline
Synonyms: 4-methyl-N,N-diphenylaniline
Molecular Weight (g/mol): 259.35
Molecular Formula: C19H17N
InChi Key: IULUNTXBHHKFFR-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results