Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyltriphenylamine 98.0+%, TCI America™
SDP

Catalog No. M18985G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18985G 5 g
M18981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18985G Supplier TCI America Supplier No. M18985G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4316-53-4
Molecular Formula C19H17N
Molecular Weight (g/mol) 259.352
InChI Key IULUNTXBHHKFFR-UHFFFAOYSA-N
Synonym N,N-Diphenyl-p-toluidine
PubChem CID 11299928
IUPAC Name 4-methyl-N,N-diphenylaniline
SMILES CC1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 67°C
Color White-Yellow
Quantity 5 g
Formula Weight 259.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyltriphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.