accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3CHNOO

CAS RN 4316-73-8

IUPAC Name: sodium 2-(methylamino)acetate
Synonyms: sarcosinate, sodium cocoyl sarcosine sodium cocoyl sarcosinate
Molecular Weight (g/mol): 111.08
Molecular Formula: C3H6NNaO2
InChi Key: ZUFONQSOSYEWCN-UHFFFAOYSA-M
SMILES: [Na+].CNCC([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results