Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sarcosine Sodium Salt (ca. 40% in Water), TCI America™
SDP

Catalog No. M025625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M025625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M025625ML Supplier TCI America Supplier No. M025625ML

Chemical Identifiers

CAS 4316-73-8
Molecular Formula C3H6NNaO2
Molecular Weight (g/mol) 111.076
MDL Number MFCD00068282
InChI Key ZUFONQSOSYEWCN-UHFFFAOYSA-M
Synonym (Methylamino)acetic Acid Sodium Salt, N-Methylglycine Sodium Salt, Sodium Sarcosinate, Sodium (Methylamino)acetate, Me-Gly-ONa
PubChem CID 23667259
IUPAC Name sodium;2-(methylamino)acetate
SMILES CNCC(=O)[O-].[Na+]

Specifications

Color Yellow
Physical Form Liquid
Quantity 25 mL
Formula Weight 111.08
Chemical Name or Material Sarcosine Sodium Salt (ca. 40% in Water)
EINECSNumber (2)-1206
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.