accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONHOCl

CAS RN 4318-56-3

IUPAC Name: 6-chloro-3-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 6-chloro-3-methyl-1H-pyrimidine-2,4-dione
Molecular Weight (g/mol): 160.56
Molecular Formula: C5H5ClN2O2
InChi Key: SGLXGFAZAARYJY-UHFFFAOYSA-N
SMILES: CN1C(=O)NC(Cl)=CC1=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)