accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNN

CAS RN 43182-10-1

IUPAC Name: 1-benzyl-1H-1,3-benzodiazol-2-amine
Synonyms: 1-benzyl-1,3-benzodiazol-2-amine
Molecular Weight (g/mol): 223.28
Molecular Formula: C14H13N3
InChi Key: LXIDSOCBAAMGJX-UHFFFAOYSA-N
SMILES: NC1=NC2=CC=CC=C2N1CC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results