Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-1-benzylbenzimidazole, 99+%, Thermo Scientific Chemicals

Catalog No. AC276290100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC276290100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC276290100 Supplier Thermo Scientific Chemicals Supplier No. 276290100

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43182-10-1
Molecular Formula C14H13N3
Molecular Weight (g/mol) 223.28
MDL Number MFCD00142854
InChI Key LXIDSOCBAAMGJX-UHFFFAOYSA-N
Synonym 2-amino-1-benzylbenzimidazole, 1-benzyl-1h-benzo d imidazol-2-amine, 1-benzyl-1h-benzimidazol-2-amine, 1-benzyl-1h-benzoimidazol-2-ylamine, 1h-benzimidazol-2-amine,1-phenylmethyl, 1-benzylbenzimidazole-2-ylamine, 1-benzyl-1,3-benzodiazol-2-amine, 2-amino-1-phenylmethyl-1h-benzimidazole, cbmicro_006128, 1-benzyl-2-aminobenzimidazole
PubChem CID 170742
SMILES NC1=NC2=CC=CC=C2N1CC1=CC=CC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 195°C to 198°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Quantity 10 g
IUPAC Name 1-benzyl-1H-1,3-benzodiazol-2-amine
Formula Weight 223.28
Percent Purity ≥99.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Amino-1-benzylbenzimidazole, 99+%
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 256-130-5
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.