accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4324-38-3

IUPAC Name: 3-ethoxypropanoic acid
Synonyms: propionic acid, ethoxy-
Molecular Weight (g/mol): 118.13
Molecular Formula: C5H10O3
InChi Key: JRXXEXVXTFEBIY-UHFFFAOYSA-N
SMILES: CCOCCC(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results