accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOOFFF

CAS RN 433-06-7

IUPAC Name: 2,2,2-trifluoroethyl 4-methylbenzene-1-sulfonate
Synonyms: 2,2,2-trifluoroethyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 254.22
Molecular Formula: C9H9F3O3S
InChi Key: IGKCQDUYZULGBM-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCC(F)(F)F
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)