accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 433-97-6

IUPAC Name: 2-(trifluoromethyl)benzoic acid
Synonyms: 2-trifluoromethylbenzoic acid α,α,α-trifluoro-o-toluic acid 2-tfmba
Molecular Weight (g/mol): 190.12
Molecular Formula: C8H5F3O2
InChi Key: FBRJYBGLCHWYOE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1C(F)(F)F
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)