Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trifluoromethyl)benzoic acid, 98%, Thermo Scientific™

Catalog No. AC188451000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC188451000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC188451000 Supplier Thermo Scientific Chemicals Supplier No. 188451000

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 433-97-6
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.12
MDL Number MFCD00002476
InChI Key FBRJYBGLCHWYOE-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzoic acid, o-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-o-toluic acid, o-carboxybenzotrifluoride, 2-carboxybenzotrifluoride, unii-0r1ck7mzua, trifluoromethylbenzoic acid, 2-trifluoromethyl benzoate, 2-trifluromethylbenzoic acid, trifluoromethyl benzoic acid
PubChem CID 9899
ChEBI CHEBI:60694
IUPAC Name 2-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=CC=CC=C1C(F)(F)F

Specifications

Melting Point 107.0°C to 110.0°C
Boiling Point 247.0°C (753.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Linear Formula CF3C6H4CO2H
Quantity 100 g
Beilstein 09,III,2308
Formula Weight 190.12
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)benzoic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 207-093-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.