accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONSHN

CAS RN 4332-97-2

IUPAC Name: 5-benzyl-3-phenyl-2-sulfanylideneimidazolidin-4-one
Synonyms: 5-benzyl-3-phenyl-2-sulfanylideneimidazolidin-4-one
Molecular Weight (g/mol): 282.36
Molecular Formula: C16H14N2OS
InChi Key: HIDCDSHFIITFOM-UHFFFAOYNA-N
SMILES: O=C1C(CC2=CC=CC=C2)NC(=S)N1C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results