Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylthiohydantoin-phenylalanine, TCI America™
SDP

Catalog No. P03671G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P03671G 1 g
P0367100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P03671G Supplier TCI America Supplier No. P03671G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4332-97-2
Molecular Formula C16H14N2OS
Molecular Weight (g/mol) 282.361
MDL Number MFCD01076623
InChI Key HIDCDSHFIITFOM-UHFFFAOYSA-N
Synonym 5-Benzyl-3-phenyl-2-thiohydantoin, PTH-phenylalanine
PubChem CID 2871911
IUPAC Name 5-benzyl-3-phenyl-2-sulfanylideneimidazolidin-4-one
SMILES C1=CC=C(C=C1)CC2C(=O)N(C(=S)N2)C3=CC=CC=C3

Specifications

Melting Point 187°C
Color White
Quantity 1 g
Formula Weight 282.36
Physical Form Crystalline Powder
Chemical Name or Material Phenylthiohydantoin-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.