accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONSHN

CAS RN 4333-22-6

IUPAC Name: 5-butyl-3-phenyl-2-sulfanylideneimidazolidin-4-one
Synonyms: 5-butyl-3-phenyl-2-sulfanylideneimidazolidin-4-one
Molecular Weight (g/mol): 248.34
Molecular Formula: C13H16N2OS
InChi Key: FJQCWUDEVCYMRZ-UHFFFAOYNA-N
SMILES: CCCCC1NC(=S)N(C1=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results