Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylthiohydantoin-norleucine 98.0+%, TCI America™
SDP

Catalog No. P0756100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0756100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0756100MG Supplier TCI America Supplier No. P0756100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4333-22-6
Molecular Formula C13H16N2OS
Molecular Weight (g/mol) 248.344
MDL Number MFCD00038437
InChI Key FJQCWUDEVCYMRZ-UHFFFAOYSA-N
Synonym 5-Butyl-3-phenyl-2-thiohydantoin, PTH-norleucine
PubChem CID 3517897
IUPAC Name 5-butyl-3-phenyl-2-sulfanylideneimidazolidin-4-one
SMILES CCCCC1C(=O)N(C(=S)N1)C2=CC=CC=C2

Specifications

Melting Point 136°C
Color White
Quantity 100 mg
Formula Weight 248.34
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phenylthiohydantoin-norleucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.