accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4335-09-5

NaNaNH2NNNOONHNNHNOOSOOOSOOO(E/Z)(E)

CAS RN 4335-09-5

IUPAC Name: disodium (3E)-5-amino-6-[2-(4-nitrophenyl)diazen-1-yl]-4-oxo-3-(2-{4'-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]-[1,1'-biphenyl]-4-yl}hydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: disodium (6E)-4-amino-3-[2-(4-nitrophenyl)diazen-1-yl]-5-oxo-6-(2-{4'-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]-[1,1'-biphenyl]-4-yl}hydrazin-1-ylidene)naphthalene-2,7-disulfonate
Molecular Weight (g/mol): 812.7
Molecular Formula: C34H22N8Na2O10S2
InChi Key: LQDBIEHCXRQVQI-LYAINCPBSA-L
SMILES: [Na+].[Na+].NC1=C(N=NC2=CC=C(C=C2)[N+]([O-])=O)C(=CC2=C1C(=O)\C(=N/NC1=CC=C(C=C1)C1=CC=C(NN=C3C=CC(=O)C=C3)C=C1)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results