Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diamine Green B, TCI America™
SDP

Catalog No. D023725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D023725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D023725G Supplier TCI America Supplier No. D023725G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4335-09-5
Molecular Formula C34H22N8Na2O10S2
Molecular Weight (g/mol) 812.70
MDL Number MFCD00042705
InChI Key LQDBIEHCXRQVQI-LYAINCPBSA-L
Synonym diamine green b, c.i. direct green 6, disodium salt, pontamine green, dianil green, 2,7-naphthalenedisulfonic acid, 4-amino-5-hydroxy-6-2-4'-2-4-hydroxyphenyl diazenyl 1,1'-biphenyl-4-yl diazenyl-3-2-4-nitrophenyl diazenyl-, sodium salt 1:2
PubChem CID 44135513
IUPAC Name disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-[[4-[4-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]phenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate
SMILES C1=CC(=O)C=CC1=NNC2=CC=C(C=C2)C3=CC=C(C=C3)NN=C4C(=CC5=CC(=C(C(=C5C4=O)N)N=NC6=CC=C(C=C6)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Black
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Diamine Green B
RTECSNumber QJ6194500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.