accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOH

CAS RN 4335-77-7

IUPAC Name: 2-cyclohexyl-2-hydroxy-2-phenylacetic acid
Synonyms: cyclohexyl(hydroxy)phenylacetic acid
Molecular Weight (g/mol): 234.3
Molecular Formula: C14H18O3
InChi Key: YTRNSQPXEDGWMR-UHFFFAOYNA-N
SMILES: OC(=O)C(O)(C1CCCCC1)C1=CC=CC=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)