Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Cyclohexyl-DL-mandelic Acid 98.0+%, TCI America™
SDP

Catalog No. C23935G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23935G 5 g
C239325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23935G Supplier TCI America Supplier No. C23935G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4335-77-7
Molecular Formula C14H18O3
Molecular Weight (g/mol) 234.295
MDL Number MFCD00194543
InChI Key YTRNSQPXEDGWMR-UHFFFAOYSA-N
Synonym alpha-cyclohexylmandelic acid, 2-cyclohexylmandelic acid, cyclohexylphenylglycolic acid, lespedamine, hexahydrobenzilic acid, cyclohexylmandelic acid, cyclohexyl-hydroxy-phenyl-acetic acid, cyclohexyl hydroxy phenylacetic acid, phenyl-cyclohexylglycolic acid, rcc 32
PubChem CID 97700
IUPAC Name 2-cyclohexyl-2-hydroxy-2-phenylacetic acid
SMILES C1CCC(CC1)C(C2=CC=CC=C2)(C(=O)O)O

Specifications

Melting Point 167°C
Color White-Yellow
Boiling Point 135°C
Quantity 5 g
Formula Weight 234.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Cyclohexyl-DL-mandelic Acid
EINECSNumber (4)-1902
RTECSNumber GV2770000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.