CAS RN 4337-33-1
CAS RN 4337-33-1
IUPAC Name:
(2-carboxyethyl)dimethylsulfanium chloride
Synonyms:
(2-carboxyethyl)dimethylsulfanium chloride
Molecular Weight (g/mol):
170.65
Molecular Formula:
C5H11ClO2S
InChi Key:
RRUMKKGRKSSZKY-UHFFFAOYSA-N
SMILES:
[Cl-].C[S+](C)CCC(O)=O
1
–
2
of
2
results
1
PESTANAL™ Dimethylpropiothetin hydrochloride, ≥96% (HPLC), MilliporeSigma™ Supelco™
Dimethylpropiothetin is a natural product found in the organisms of saline environment such as algae and aquatic plants.
2
(2-Carboxyethyl)dimethylsulfonium Chloride 98.0+%, TCI America™
CAS: 4337-33-1 Molecular Formula: C5H11ClO2S Molecular Weight (g/mol): 170.65 MDL Number: MFCD00142888 InChI Key: RRUMKKGRKSSZKY-UHFFFAOYSA-N Synonym: Dimethyl-beta-propiothetin PubChem CID: 5316899 IUPAC Name: (2-carboxyethyl)dimethylsulfanium chloride SMILES: [Cl-].C[S+](C)CCC(O)=O