Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Carboxyethyl)dimethylsulfonium Chloride 98.0+%, TCI America™
SDP

Catalog No. C12405G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C12405G Supplier TCI America Supplier No. C12405G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4337-33-1
Molecular Formula C5H11ClO2S
Molecular Weight (g/mol) 170.65
MDL Number MFCD00142888
InChI Key RRUMKKGRKSSZKY-UHFFFAOYSA-N
Synonym Dimethyl-beta-propiothetin
PubChem CID 5316899
IUPAC Name (2-carboxyethyl)dimethylsulfanium chloride
SMILES [Cl-].C[S+](C)CCC(O)=O

Specifications

Melting Point 128°C
Color White
Quantity 5 g
Formula Weight 170.65
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (2-Carboxyethyl)dimethylsulfonium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.