accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHNBr

CAS RN 433711-95-6

IUPAC Name: tert-butyl N-(2-bromopyridin-4-yl)carbamate
Synonyms: tert-butyl N-(2-bromopyridin-4-yl)carbamate
Molecular Weight (g/mol): 273.13
Molecular Formula: C10H13BrN2O2
InChi Key: DCYAZECOQNZWBD-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC=NC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results