Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Boc-amino)-2-bromopyridine, 97%, Thermo Scientific™

Catalog No. AAL1984703
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1984703 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1984703 Supplier Thermo Scientific Chemicals Supplier No. L1984703

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 433711-95-6
Molecular Formula C10H13BrN2O2
Quantity 1g
MDL Number MFCD03791266
Synonym 2-bromo-4-boc pyridine, 4-boc-amino-2-bromopyridine, tert-butyl 2-bromopyridin-4-ylcarbamate, tert-butyl 2-bromopyridin-4-yl carbamate, 2-bromo-pyridin-4-yl carbamic acid tert-butyl ester, tert-butyl n-2-bromopyridin-4-yl carbamate, 2-bromo-4-n-boc-aminopyridine, 2-bromo-4-tert-butoxycarbonylamino pyridine, tert-butoxy-n-2-bromo 4-pyridyl carboxamide, tert-butyl2-bromopyridin-4-ylcarbamate
InChI Key DCYAZECOQNZWBD-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)NC1=CC=NC(Br)=C1
IUPAC Name tert-butyl N-(2-bromopyridin-4-yl)carbamate
Molecular Weight (g/mol) 273.13
PubChem CID 7015522
Formula Weight 273.13
Percent Purity 97%
Chemical Name or Material 4-(Boc-amino)-2-bromopyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.