accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHO

CAS RN 4342-60-3

IUPAC Name: 4-methylcyclohex-3-ene-1-carboxylic acid
Synonyms: 4-methylcyclohex-3-ene-1-carboxylic acid
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H12O2
InChi Key: OYOQOLNBTPTFEM-UHFFFAOYNA-N
SMILES: CC1=CCC(CC1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results