Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-3-cyclohexene-1-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. M24925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24925G 5 g
M24921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24925G Supplier TCI America Supplier No. M24925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4342-60-3
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.182
InChI Key OYOQOLNBTPTFEM-UHFFFAOYSA-N
PubChem CID 20350
IUPAC Name 4-methylcyclohex-3-ene-1-carboxylic acid
SMILES CC1=CCC(CC1)C(=O)O

Specifications

Melting Point 100°C
Color White
Quantity 5 g
Formula Weight 140.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-3-cyclohexene-1-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.