CAS RN 4349-07-9
IUPAC Name:
5-iodo-3,4-dihydropyrimidin-4-one
Synonyms:
5-iodo-3H-pyrimidin-4-one
Molecular Weight (g/mol):
221.99
Molecular Formula:
C4H3IN2O
InChi Key:
SMIGOGPUTQXOKF-UHFFFAOYSA-N
SMILES:
IC1=CN=CNC1=O
1
–
1
of
1
results
1