accessibility menu, dialog, popup

UserName

Viewing profile as user:
FONNH

CAS RN 436-72-6

IUPAC Name: 5-fluoro-1,4-dihydroquinazolin-4-one
Synonyms: 5-fluoro-1H-quinazolin-4-one
Molecular Weight (g/mol): 164.14
Molecular Formula: C8H5FN2O
InChi Key: UXEZULVIMJVIFB-UHFFFAOYSA-N
SMILES: FC1=C2C(NC=NC2=O)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results