CAS RN 4362-40-7
CAS RN 4362-40-7
IUPAC Name:
4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane
Synonyms:
4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane
Molecular Weight (g/mol):
150.6
Molecular Formula:
C6H11ClO2
InChi Key:
BNPOTXLWPZOESZ-UHFFFAOYNA-N
SMILES:
CC1(C)OCC(CCl)O1
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich 4-Chloromethyl-2,2-dimethyl-1,3-dioxolane
Sigma Aldrich 4-Chloromethyl-2,2-dimethyl-1,3-dioxolane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.