accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHOONOO

CAS RN 4368-83-6

IUPAC Name: 5-acetamido-2-nitrobenzoate
Synonyms: 5-acetamido-2-nitrobenzoate
Molecular Weight (g/mol): 223.17
Molecular Formula: C9H7N2O5
InChi Key: ZSHFMOUMOUOGKI-UHFFFAOYSA-M
SMILES: CC(=O)NC1=CC=C(C(=C1)C([O-])=O)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results