Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Acetamido-2-nitrobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. A21565G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A21565G Supplier TCI America Supplier No. A21565G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4368-83-6
Molecular Formula C9H8N2O5
Molecular Weight (g/mol) 224.172
MDL Number MFCD00035889
InChI Key ZSHFMOUMOUOGKI-UHFFFAOYSA-N
Synonym 5-acetamido-2-nitrobenzoic acid, 5-acetylamino-2-nitrobenzoic acid, benzoic acid, 5-acetylamino-2-nitro, 2-nitro-5-acetylaminobenzoic acid, 5-acetamido-2-nitrobenzoicacid, pubchem13220, acmc-209jv7, 5-acetamido-2-nitrobenzoic
PubChem CID 78076
ChEBI CHEBI:86569
SMILES CC(=O)NC1=CC(=C(C=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 225°C
Color Yellow
Quantity 5 g
Formula Weight 224.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Acetamido-2-nitrobenzoic Acid
EINECSNumber (3)-1513
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.