accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4369-55-5

IUPAC Name: 3-phenyl-1,2-oxazol-5-amine
Synonyms: 3-phenyl-1,2-oxazol-5-amine
Molecular Weight (g/mol): 160.18
Molecular Formula: C9H8N2O
InChi Key: HLOHVVZZMMMDMM-UHFFFAOYSA-N
SMILES: NC1=CC(=NO1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results