CAS RN 437-13-8
IUPAC Name:
tetrafluoroboranuide; triphenylsulfanium
Synonyms:
triphenylsulfonium tetrafluoroborate
Molecular Weight (g/mol):
350.18
Molecular Formula:
C18H15BF4S
InChi Key:
RTWMEMGVYYTCOZ-UHFFFAOYSA-N
SMILES:
F[B-](F)(F)F.C1=CC=C(C=C1)[S+](C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Triphenylsulfonium Tetrafluoroborate 98.0+%, TCI America™
CAS: 437-13-8 Molecular Formula: C18H15BF4S Molecular Weight (g/mol): 350.18 MDL Number: MFCD00671581 InChI Key: RTWMEMGVYYTCOZ-UHFFFAOYSA-N PubChem CID: 2760877 IUPAC Name: tetrafluoroboranuide; triphenylsulfanium SMILES: F[B-](F)(F)F.C1=CC=C(C=C1)[S+](C1=CC=CC=C1)C1=CC=CC=C1