accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 437-86-5

IUPAC Name: 2-fluoro-1-methyl-3-nitrobenzene
Synonyms: 2-fluoro-1-methyl-3-nitrobenzene
Molecular Weight (g/mol): 155.13
Molecular Formula: C7H6FNO2
InChi Key: NBCNUIXYBLFJMI-UHFFFAOYSA-N
SMILES: CC1=C(F)C(=CC=C1)[N+]([O-])=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)