accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOOHOHOOOH

CAS RN 4371-28-2

IUPAC Name: [1,1'-biphenyl]-3,3',5,5'-tetracarboxylic acid
Synonyms: [1,1'-biphenyl]-3,3',5,5'-tetracarboxylic acid
Molecular Weight (g/mol): 330.25
Molecular Formula: C16H10O8
InChi Key: QURGMSIQFRADOZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC(=C1)C(O)=O)C1=CC(=CC(=C1)C(O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)