Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biphenyl-3,3',5,5'-tetracarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. B3792200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3792200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B3792200MG Supplier TCI America Supplier No. B3792200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4371-28-2
Molecular Formula C16H10O8
Molecular Weight (g/mol) 330.248
MDL Number MFCD16621475
InChI Key QURGMSIQFRADOZ-UHFFFAOYSA-N
PubChem CID 15840397
IUPAC Name 5-(3,5-dicarboxyphenyl)benzene-1,3-dicarboxylic acid
SMILES C1=C(C=C(C=C1C(=O)O)C(=O)O)C2=CC(=CC(=C2)C(=O)O)C(=O)O

Specifications

Color White
Quantity 200 mg
Formula Weight 330.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Biphenyl-3,3′,5,5′-tetracarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.