accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)(S)OHNH2OOH3CCH3CH3

CAS RN 4378-13-6

IUPAC Name: (2S,3R)-2-amino-3-(tert-butoxy)butanoic acid
Synonyms: O-tert-butyl-L-threonine
Molecular Weight (g/mol): 175.23
Molecular Formula: C8H17NO3
InChi Key: NMJINEMBBQVPGY-RITPCOANSA-N
SMILES: C[C@@H](OC(C)(C)C)[C@H](N)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)