Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Nitronium Tetrafluoroborate Solution

Catalog No. 176303200ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-630-3200ML 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 17-630-3200ML Supplier Sigma Aldrich Supplier No. 176303200ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for preparation of:Anologues of SCH 39166 as potent and selective dopamine D1 receptor antagonistsAnticonvulsant agent SB-406725A Nitro-derivatives of chlorambucil conjugated to prasterone and pregnenolone as anticancer agentsNano-sized cryptomelane particles for the formation of potentiometric potassium ion sensorsNitro- and aminoindole derivativesHydroxynitroalkyl esters as affectors of metabolism of sulfate-reducing bacteriaFluorescence probes to monitor the generation of nitrative stress in neurodegenerative disordersUsed as:Oxidizing agent in high-power Li-ion battery technology for large-scale applications as required for plug-in hybrid vehicles

Specifications

CAS 4378-13-6
Molecular Formula Pd
Linear Formula Pd
MDL Number MFCD03457879
Quantity 25 g
Synonym Pd/C
Molecular Weight (g/mol) 106.42 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.