accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4378-43-2

IUPAC Name: 4-phosphonatobutanoate
Synonyms: 4-phosphonatobutanoate
Molecular Weight (g/mol): 165.06
Molecular Formula: C4H6O5P
InChi Key: UYRFPODVSLYSCO-UHFFFAOYSA-K
SMILES: [O-]C(=O)CCCP([O-])([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results