Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phosphonobutyric Acid 98.0+%, TCI America™
SDP

Catalog No. P2349200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2349200MG 200 mg
P23491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2349200MG Supplier TCI America Supplier No. P2349200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4378-43-2
Molecular Formula C4H9O5P
Molecular Weight (g/mol) 168.085
MDL Number MFCD00041414
InChI Key UYRFPODVSLYSCO-UHFFFAOYSA-N
Synonym (3-Carboxypropyl)phosphonic Acid
PubChem CID 2773805
IUPAC Name 4-phosphonobutanoic acid
SMILES C(CC(=O)O)CP(=O)(O)O

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 200 mg
UN Number 3261
Formula Weight 168.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phosphonobutyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.