accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)(S)(S)(R)(S)H3C(S)

CAS RN 438-22-2

IUPAC Name: (3aS,3bS,5aR,9aS,9bS,11aS)-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthrene
Synonyms: 5α-androstane etioallocholane aetioallocholane
Molecular Weight (g/mol): 260.47
Molecular Formula: C19H32
InChi Key: QZLYKIGBANMMBK-UGCZWRCOSA-N
SMILES: C[C@@]12CCC[C@H]1[C@@H]1CC[C@H]3CCCC[C@]3(C)[C@H]1CC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results