Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-a-Androstane Solution, 2000μg/mL in Dichloromethane, Ultra Scientific

Catalog No. USIST5001
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-IST-500-1 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-IST-500-1 Supplier Agilent Technologies Supplier No. IST5001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 438-22-2 , 75-09-2
Molecular Formula C19H32
Molecular Weight (g/mol) 260.465
InChI Key QZLYKIGBANMMBK-UGCZWRCOSA-N
PubChem CID 94144
ChEBI CHEBI:28859
IUPAC Name (5R,8S,9S,10S,13S,14S)-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene
SMILES CC12CCCC1C3CCC4CCCCC4(C3CC2)C

Specifications

CAS Max % 99.85, 0.15
Physical Form Liquid
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2000μg/mL
Chemical Name or Material 5-a-Androstane Solution, 2000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.