accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4383-06-6

IUPAC Name: 5-(hydroxymethyl)-2-methoxyphenol
Synonyms: 5-(hydroxymethyl)-2-methoxyphenol
Molecular Weight (g/mol): 154.17
Molecular Formula: C8H10O3
InChi Key: WHKRHBLAJFYZKF-UHFFFAOYSA-N
SMILES: COC1=CC=C(CO)C=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results