Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isovanillyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. I06485G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I06485G Supplier TCI America Supplier No. I06485G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4383-06-6
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
InChI Key WHKRHBLAJFYZKF-UHFFFAOYSA-N
Synonym 3-hydroxy-4-methoxybenzyl alcohol, 5-hydroxymethyl-2-methoxyphenol, isovanillyl alcohol, 3-hydroxy-4-methoxybenzylalcohol, benzenemethanol, 3-hydroxy-4-methoxy, 4-methoxy-3-hydroxy benzyl alcohol, pubchem15050, ksc495i5l, 3-hydroxy-4-methoxy-benzyl alcohol, 3-hydroxy-4-methoxy benzyl alcohol
PubChem CID 78089
IUPAC Name 5-(hydroxymethyl)-2-methoxyphenol
SMILES COC1=C(C=C(C=C1)CO)O

Specifications

Melting Point 136°C
Color White
Quantity 5 g
Formula Weight 154.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Isovanillyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.