CAS RN 4383-22-6
IUPAC Name:
benzyl(prop-2-en-1-yl)amine
Synonyms:
benzyl(prop-2-en-1-yl)amine
Molecular Weight (g/mol):
147.22
Molecular Formula:
C10H13N
InChi Key:
RHUCQDQRNUUMKY-UHFFFAOYSA-N
SMILES:
C=CCNCC1=CC=CC=C1
1
–
1
of
1
results
1
N-Allylbenzylamine 97.0+%, TCI America™
CAS: 4383-22-6 Molecular Formula: C10H13N Molecular Weight (g/mol): 147.22 MDL Number: MFCD00463446 InChI Key: RHUCQDQRNUUMKY-UHFFFAOYSA-N Synonym: 3-(Benzylamino)-1-propene, N-Benzylallylamine PubChem CID: 521150 IUPAC Name: benzyl(prop-2-en-1-yl)amine SMILES: C=CCNCC1=CC=CC=C1