accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4385-35-7

IUPAC Name: 3,4-dihydro-1H-2-benzopyran-3-one
Synonyms: 3-isochromanone
Molecular Weight (g/mol): 148.16
Molecular Formula: C9H8O2
InChi Key: ILHLUZUMRJQEAH-UHFFFAOYSA-N
SMILES: O=C1CC2=CC=CC=C2CO1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)